Methyl 3-bromo-5-fluoropyridine-2-carboxylate 96%

Supplier: Ambeed
Warning

A337910-1G A337910-250MG
76989-378EA 75 USD
76989-378 77885-890
Methyl 3-bromo-5-fluoropyridine-2-carboxylate 96%
Methyl 3-bromo-5-fluoropyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₇H₅BrFNO₂
MDL Number: MFCD14698117
CAS Number: 1214337-00-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR