4-[(Pyridin-4-yl)oxy]benzenesulfonyl chloride hydrochloride 97%

Supplier: Ambeed
Danger

Synonyms: 4-(Pyridin-4-yloxy)benzene-1-sulphonyl chloride hydrochloride

A337743-100MG A337743-1G A337743-250MG
77033-002EA 41.38 USD
77033-002 77033-004 77033-006
4-[(Pyridin-4-yl)oxy]benzenesulfonyl chloride hydrochloride 97%
4-[(Pyridin-4-yl)oxy]benzenesulfonyl chloride hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₈ClNO₃S·ClH
MW: 306.165 g/mol
MDL Number: MFCD01631897
CAS Number: 192330-49-7
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR