Boc-D-Ser(tBu)-OH·DCHA 97%

Supplier: Ambeed
Warning

Synonyms: Boc-D-Ser(tBu)-OH.DCHA, Boc-D-Ser(tBu)-OH·DCHA

A337670-1G A337670-250MG
77003-086EA 21 USD
77003-086 77003-088
Boc-D-Ser(tBu)-OH·DCHA 97%
Boc-D-Ser(tBu)-OH·DCHA

Precaución: for Research Use Only

Formula: C₂₄H₄₆N₂O₅
MW: 442.641 g/mol
MDL Number: MFCD00236872
CAS Number: 248921-67-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR