5-Bromo-2-methoxy-4-methylbenzoic acid 95%

Supplier: Ambeed
Warning

A337540-100MG A337540-250MG
76989-368EA 37.14 USD
76989-368 76989-370
5-Bromo-2-methoxy-4-methylbenzoic acid 95%
5-Bromo-2-methoxy-4-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉BrO₃
MW: 245.07 g/mol
MDL Number: MFCD21234732
CAS Number: 90326-61-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR