2-Amino-4-(trifluoromethyl)pyrimidine 97%

Supplier: Ambeed
Warning

A336709-10G A336709-1G A336709-25G A336709-5G
77015-496EA 26.53 USD
77015-496 77015-498 77015-500 77015-502
2-Amino-4-(trifluoromethyl)pyrimidine 97%
2-Amino-4-(trifluoromethyl)pyrimidine

Precaución: for Research Use Only

Formula: C₅H₄F₃N₃
MW: 163.1 g/mol
MDL Number: MFCD00828624
CAS Number: 16075-42-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR