1,4-Benzenediboronic acid bis(neopentyl glycol) ester 98%

Supplier: Ambeed
Warning

Synonyms: 1,4-Bis(5,5-dimethyl-1,3,2-dioxaborinan-2-yl)benzene

A336491-1G A336491-250MG A336491-5G
77032-994EA 28 USD
77032-994 77032-996 77872-491
1,4-Benzenediboronic acid bis(neopentyl glycol) ester 98%
1,4-Benzenediboronic acid bis(neopentyl glycol) ester

Precaución: for Research Use Only

Formula: C₁₆H₂₄B₂O₄
MDL Number: MFCD01075726
CAS Number: 5565-36-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR