2,4,6-Trifluoroanisole 95+%

Supplier: Ambeed
Danger

A336345-10G A336345-250MG A336345-5G
77003-074EA 185 USD
77003-074 77003-076 77003-078
2,4,6-Trifluoroanisole 95+%
2,4,6-Trifluoroanisole

Precaución: for Research Use Only

Formula: C₇H₅F₃O
MDL Number: MFCD01631502
CAS Number: 219998-30-8
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR