1-Bromo-2,3-difluoro-5-nitrobenzene 95%

Supplier: Ambeed
Warning

A336247-100MG A336247-1G A336247-250MG A336247-5G
77015-486EA 11 USD
77015-486 77015-488 77015-490 77015-492
1-Bromo-2,3-difluoro-5-nitrobenzene 95%
1-Bromo-2,3-difluoro-5-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrF₂NO₂
MDL Number: MFCD03095339
CAS Number: 374633-24-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR