2-(3-Bromophenyl)acetamide 96%

Supplier: Ambeed
Warning

A336219-250MG A336219-5G
76989-344EA 15 USD
76989-344 76989-346
2-(3-Bromophenyl)acetamide 96%
2-(3-Bromophenyl)acetamide

Precaución: for Research Use Only

Formula: C₈H₈BrNO
MDL Number: MFCD07781225
CAS Number: 60312-83-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR