N,N,N',N'-Bis(tetramethylene)-O-(N-succinimidyl)uronium hexafluorophosphate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-(((2,5-Dioxopyrrolidin-1-yl)oxy)(pyrrolidin-1-yl)methylene)pyrrolidin-1-ium hexafluorophosphate(V), HSPyU, Dipyrrolidino(N-succinimidyloxy)carbenium Hexafluorophosphate, Dipyrrolidino(N-succinimidyloxy)carbenium hexafluorophosphate
A334790-1G
A334790-250MG
A334790-25G
A334790-5G
77003-048EA
27.99
USD
77003-048
77003-050
77003-052
77003-054
N,N,N',N'-Bis(tetramethylene)-O-(N-succinimidyl)uronium hexafluorophosphate 98%
N,N,N',N'-Bis(tetramethylene)-O-(N-succinimidyl)uronium hexafluorophosphate
Precaución: for Research Use Only
Formula:
C₁₃H₂₀F₆N₃O₃P |
MDL Number:
MFCD00192457 CAS Number: 207683-26-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...