(2S,4R)-1-(tert-Butoxycarbonyl)-4-(2-iodobenzyl)pyrrolidine-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A334723-100MG A334723-250MG
76989-322EA 101.85 USD
76989-322 76989-324
(2S,4R)-1-(tert-Butoxycarbonyl)-4-(2-iodobenzyl)pyrrolidine-2-carboxylic acid 97%
(2S,4R)-1-(tert-Butoxycarbonyl)-4-(2-iodobenzyl)pyrrolidine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₇H₂₂INO₄
MW: 431.27 g/mol
MDL Number: MFCD06659436
CAS Number: 959573-29-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR