L(-)-Methioninol 97%

Supplier: Ambeed
Warning

A334709-10G A334709-5G A334709-1G A334709-25G
77003-044EA 52 USD
77003-044 77003-046 77881-924 77881-925
L(-)-Methioninol 97%
L(-)-Methioninol

Precaución: for Research Use Only

Formula: C₅H₁₃NOS
MDL Number: MFCD00004735
CAS Number: 2899-37-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR