2-(Bis(2-(tert-butoxy)-2-oxoethyl)amino)acetic acid 97%

Supplier: Ambeed
Warning

A334350-10G A334350-1G A334350-250MG
76989-316EA 253.56 USD
76989-316 76989-318 76989-320
2-(Bis(2-(tert-butoxy)-2-oxoethyl)amino)acetic acid 97%
2-(Bis(2-(tert-butoxy)-2-oxoethyl)amino)acetic acid

Precaución: for Research Use Only

Formula: C₁₄H₂₅NO₆
MW: 303.35 g/mol
MDL Number: MFCD09833730
CAS Number: 171557-31-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR