Methyl 4-amino-3-cyanobenzoate ≥97%
Supplier: Ambeed
|
Warning
|
A334241-100MG
A334241-10G
A334241-1G
A334241-250MG
A334241-5G
77015-452EA
43.5
USD
77015-452
77015-454
77015-456
77015-458
77683-998
Methyl 4-amino-3-cyanobenzoate ≥97%
Methyl 4-amino-3-cyanobenzoate
Precaución: For research use only.
Formula:
C₉H₈N₂O₂ MW: 176.172 g/mol |
MDL Number:
MFCD16038776 CAS Number: 159847-80-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...