tert-Butyl (1-amino-2-methylpropan-2-yl)carbamate 97%
Supplier: Ambeed
|
Warning
|
A334171-100MG
A334171-1G
A334171-250MG
76989-310EA
32.89
USD
76989-310
77550-952
77683-997
tert-Butyl (1-amino-2-methylpropan-2-yl)carbamate 97%
tert-Butyl (1-amino-2-methylpropan-2-yl)carbamate
Precaución: For research use only.
Formula:
C₉H₂₀N₂O₂ MW: 188.27 g/mol |
MDL Number:
MFCD14529625 CAS Number: 320581-09-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...