6-Chloro-5-nitro-1H-indazole 98%

Supplier: Ambeed
Warning

A334100-100MG A334100-250MG A334100-5G A334100-1G A334100-25G
76989-306EA 28.65 USD
76989-306 76989-308 77593-972 77612-914 77612-916
6-Chloro-5-nitro-1H-indazole 98%
6-Chloro-5-nitro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄ClN₃O₂
MW: 197.58 g/mol
MDL Number: MFCD08690109
CAS Number: 101420-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR