Benzofuran-4-ol 95+%

Supplier: Ambeed
Warning

A333901-100MG A333901-250MG
76989-300EA 106 USD
76989-300 76989-302
Benzofuran-4-ol 95+%
Benzofuran-4-ol

Precaución: for Research Use Only

Formula: C₈H₆O₂
MDL Number: MFCD09754184
CAS Number: 480-97-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR