1-(3-Isopropoxyphenyl)ethanone 98%

Supplier: Ambeed
Warning

A333593-1G A333593-5G
77032-956EA 43.5 USD
77032-956 77032-958
1-(3-Isopropoxyphenyl)ethanone 98%
1-(3-Isopropoxyphenyl)ethanone

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₂
MDL Number: MFCD01922015
CAS Number: 114590-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR