EPPS (HEPPS, 4-(2-Hydroxyethyl)-1-piperazinepropanesulfonic acid) 98%

Supplier: Ambeed
Warning

Synonyms: 4-(2-Hydroxyethyl)-1-piperazinepropanesulphonic acid, N-(2-Hydroxyethyl)piperazine-N′-(3-propanesulphonic acid), HEPPS, 3-[4-(2-Hydroxyethyl)-1-piperazinyl]propanesulphonic acid, 3-[4-(2-Hydroxyethyl)-1-piperazinyl]-propanesulphonic acid, 4-(2-Hydroxyethyl)piperazine-1-propanesulphonic acid

A333394-100G A333394-25G A333394-500G
77032-926EA 38.2 USD
77032-926 77032-928 77032-930
EPPS (HEPPS, 4-(2-Hydroxyethyl)-1-piperazinepropanesulfonic acid) 98%
EPPS (HEPPS, 4-(2-hydroxyethyl)-1-piperazinepropanesulphonic acid)

Precaución: for Research Use Only

Formula: C₉H₂₀N₂O₄S
MW: 252.33 g/mol
MDL Number: MFCD00006160
CAS Number: 16052-06-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR