7-Azaindole-3-boronic Acid Pinacol Ester 96%

Supplier: Ambeed
Warning

A332512-1G A332512-100MG A332512-250MG
77032-902EA 143 USD
77032-902 77872-479 77872-480
7-Azaindole-3-boronic Acid Pinacol Ester 96%
7-Azaindole-3-boronic Acid Pinacol Ester

Precaución: for Research Use Only

Formula: C₁₃H₁₇BN₂O₂
MW: 244.09 g/mol
MDL Number: MFCD11616359
CAS Number: 945256-29-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR