tert-Butyl (2-aminoethyl)(ethyl)carbamate 95%

Supplier: Ambeed
Warning

Synonyms: (2-Amino-ethyl)-ethyl-carbamic acid tert-butyl ester

A332390-100MG A332390-1G A332390-250MG A332390-5G A332390-25G
77015-434EA 27.59 USD
77015-434 77015-436 77015-438 77015-440 77683-991
tert-Butyl (2-aminoethyl)(ethyl)carbamate 95%
tert-Butyl (2-aminoethyl)(ethyl)carbamate

Precaución: For research use only.

Formula: C₉H₂₀N₂O₂
MW: 188.267 g/mol
MDL Number: MFCD04114276
CAS Number: 105628-63-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR