Rosiglitazone maleate 98%

Supplier: Ambeed
Warning

Synonyms: 5-(4-(2-(Methyl(pyridin-2-yl)amino)ethoxy)benzyl)thiazolidine-2,4-dione maleate

A332359-10G A332359-10MG A332359-1G A332359-250MG A332359-50MG A332359-5G
77003-028EA 9.55 USD
77003-028 77003-030 77003-032 77003-034 77003-036 77003-038 77003-040
Rosiglitazone maleate 98%
Rosiglitazone maleate

Precaución: for Research Use Only

Formula: C₁₈H₁₉N₃O₃S·C₄H₄O₄
MW: 473.4989 g/mol
MDL Number: MFCD03427306
CAS Number: 155141-29-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR