4-Methoxyquinolin-8-amine 95%

Supplier: Ambeed
Warning

A332207-1G A332207-250MG
77015-430EA 342.68 USD
77015-430 77015-432
4-Methoxyquinolin-8-amine 95%
4-Methoxyquinolin-8-amine

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O
MW: 174.2 g/mol
MDL Number: MFCD09261105
CAS Number: 59665-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR