1-Bromo-2,3-dichloro-4-fluorobenzene 98%

Supplier: Ambeed
Warning

A332179-1G A332179-5G
76989-280EA 44.56 USD
76989-280 76989-282
1-Bromo-2,3-dichloro-4-fluorobenzene 98%
1-Bromo-2,3-dichloro-4-fluorobenzene

Precaución: for Research Use Only

Formula: C₆H₂BrCl₂F
MW: 243.89 g/mol
MDL Number: MFCD11845895
CAS Number: 1093092-14-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR