2,4-Bis(2,4-dimethylphenyl)-6-(2-hydroxy-4-n-octyloxyphenyl)-1,3,5-triazine 98%

Supplier: Ambeed
Warning

A331174-100G A331174-25G A331174-500G A331174-5G
77003-020EA 35 USD
77003-020 77003-022 77003-024 77003-026
2,4-Bis(2,4-dimethylphenyl)-6-(2-hydroxy-4-n-octyloxyphenyl)-1,3,5-triazine 98%
2,4-Bis(2,4-dimethylphenyl)-6-(2-hydroxy-4-n-octyloxyphenyl)-1,3,5-triazine

Precaución: for Research Use Only

Formula: C₃₃H₃₉N₃O₂
MDL Number: MFCD09834146
CAS Number: 2725-22-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR