N-α-Benzoyl-L-arginine 98,5%

Supplier: Ambeed
Warning

A330648-100G A330648-25G A330648-5G
77003-014EA 62.99 USD
77003-014 77003-016 77003-018
N-α-Benzoyl-L-arginine 98,5%
N-alpha-Benzoyl-L-arginine

Precaución: for Research Use Only

Formula: C₁₃H₁₈N₄O₃
MDL Number: MFCD00001763
CAS Number: 154-92-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR