2,3,6-Trifluorocinnamic acid 95%

Supplier: Ambeed
Warning

A330152-1G A330152-5G
77003-010EA 43 USD
77003-010 77003-012
2,3,6-Trifluorocinnamic acid 95%
2,3,6-Trifluorocinnamic acid

Precaución: for Research Use Only

Formula: C₉H₅F₃O₂
MDL Number: MFCD00061213
CAS Number: 230295-12-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR