2-(Pyridin-4-yloxy)acetic acid 95%

Supplier: Ambeed
Warning

A330072-100MG A330072-250MG A330072-5G
76989-264EA 122.01 USD
76989-264 76989-266 76989-268
2-(Pyridin-4-yloxy)acetic acid 95%
2-(Pyridin-4-yloxy)acetic acid

Precaución: for Research Use Only

Formula: C₇H₇NO₃
MW: 153.14 g/mol
MDL Number: MFCD17170979
CAS Number: 58530-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR