5-Bromo-7-methyl-1H-indazole 97%

Supplier: Ambeed
Warning

A330017-100G A330017-10G A330017-1G A330017-250MG A330017-25G A330017-5G
77015-404EA 1092.73 USD
77015-404 77015-406 77015-408 77015-410 77015-412 77015-414
5-Bromo-7-methyl-1H-indazole 97%
5-Bromo-7-methyl-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₇BrN₂
MW: 211.0600 g/mol
MDL Number: MFCD03990484
CAS Number: 156454-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR