4-(4-Aminophenoxy)pyridin-2(1H)-one 95%

Supplier: Ambeed
Warning

A329908-1G A329908-5G A329908-250MG
76989-260EA 434.97 USD
76989-260 76989-262 77683-985
4-(4-Aminophenoxy)pyridin-2(1H)-one 95%
4-(4-Aminophenoxy)pyridin-2(1H)-one

Precaución: For research use only.

Formula: C₁₁H₁₀N₂O₂
MW: 202.21 g/mol
MDL Number: MFCD24038953
CAS Number: 1041861-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR