2-Acetyl-5,5-dimethyl-1,3-cyclohexanedione 98%

Supplier: Ambeed
Warning

A329749-1G A329749-250MG A329749-5G
76989-254EA 18 USD
76989-254 76989-256 76989-258
2-Acetyl-5,5-dimethyl-1,3-cyclohexanedione 98%
2-Acetyl-5,5-dimethyl-1,3-cyclohexanedione

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₃
MDL Number: MFCD00467663
CAS Number: 1755-15-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR