4-Chloro-3,5-difluorobenzoic acid 97%

Supplier: Ambeed
Warning

A329681-1G A329681-25G A329681-5G
77015-398EA 24.41 USD
77015-398 77015-400 77015-402
4-Chloro-3,5-difluorobenzoic acid 97%
4-Chloro-3,5-difluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃ClF₂O₂
MW: 192.547 g/mol
MDL Number: MFCD11554154
CAS Number: 1160573-19-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR