5-Fluoro-2-methoxy-4-pyrimidinone 97%

Supplier: Ambeed
Warning

A329577-100G A329577-10G A329577-25G A329577-5G
77015-392EA 22 USD
77015-392 77015-394 77015-396 77890-655
5-Fluoro-2-methoxy-4-pyrimidinone 97%
5-Fluoro-2-methoxy-4-pyrimidinone

Precaución: for Research Use Only

Formula: C₅H₅FN₂O₂
MDL Number: MFCD03265437
CAS Number: 1480-96-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR