5,6,12,13-Tetrachloroanthra[2,1,9-def:6,5,10-d'e'f']diisochromene-1,3,8,10-tetraone 95%
Supplier: Ambeed
|
Warning
|
A329223-100G
A329223-10G
A329223-1G
A329223-25G
A329223-5G
A329223-250MG
77002-994EA
165
USD
77002-994
77002-996
77002-998
77003-000
77003-002
77672-690
5,6,12,13-Tetrachloroanthra[2,1,9-def:6,5,10-d'e'f']diisochromene-1,3,8,10-tetraone 95%
5,6,12,13-Tetrachloroanthra[2,1,9-def:6,5,10-d'e'f']diisochromene-1,3,8,10-tetraone
Precaución: for Research Use Only
Formula:
C₂₄H₄Cl₄O₆ |
MDL Number:
MFCD08056065 CAS Number: 156028-26-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...