2-Hydrazinyl-4-methylbenzo[d]thiazole 95%

Supplier: Ambeed
Warning

A329208-100G A329208-25G
77002-990EA 37.14 USD
77002-990 77002-992
2-Hydrazinyl-4-methylbenzo[d]thiazole 95%
2-Hydrazinyl-4-methylbenzo[d]thiazole

Precaución: for Research Use Only

Formula: C₈H₉N₃S
MW: 179.24 g/mol
MDL Number: MFCD04448793
CAS Number: 20174-68-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR