3-Bromo-5-fluoro-2-methylpyridine 98%
Supplier: Ambeed
|
Warning
|
A329032-1G
A329032-5G
A329032-100MG
A329032-250MG
76989-240EA
105.03
USD
76989-240
77550-918
77683-981
77683-982
3-Bromo-5-fluoro-2-methylpyridine 98%
3-Bromo-5-fluoro-2-methylpyridine
Precaución: For research use only.
Formula:
C₆H₅BrFN |
MDL Number:
MFCD18255300 CAS Number: 1211542-29-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...