(1-Methylpiperidin-2-yl)methanamine dihydrochloride 95%

Supplier: Ambeed
Warning

A328760-1G A328760-250MG A328760-5G
77015-386EA 455 USD
77015-386 77015-388 77015-390
(1-Methylpiperidin-2-yl)methanamine dihydrochloride 95%
(1-Methylpiperidin-2-yl)methanamine dihydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₈Cl₂N₂
MDL Number: MFCD27996802
CAS Number: 1440799-70-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR