2,6-Dichloro-4-methoxyaniline 95%

Supplier: Ambeed
Warning

A328754-100MG A328754-10G A328754-1G A328754-250MG A328754-5G
77015-376EA 22 USD
77015-376 77015-378 77015-380 77015-382 77015-384
2,6-Dichloro-4-methoxyaniline 95%
2,6-Dichloro-4-methoxyaniline

Precaución: for Research Use Only

Formula: C₇H₇Cl₂NO
MDL Number: MFCD00115065
CAS Number: 6480-66-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR