2,6-Dichloro-5-methylpyridin-3-amine 95%

Supplier: Ambeed
Danger

A328670-100MG A328670-1G A328670-250MG
76989-232EA 8 USD
76989-232 76989-234 76989-236
2,6-Dichloro-5-methylpyridin-3-amine 95%
2,6-Dichloro-5-methylpyridin-3-amine

Precaución: for Research Use Only

Formula: C₆H₆Cl₂N₂
MDL Number: MFCD20697565
CAS Number: 58596-89-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR