(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)decanoic acid 95%

Supplier: Ambeed
Warning

Synonyms: (S)-N-FMOC-Octylglycine, N-Fmoc-2-octyl-L-glycine, Fmoc-octyl-Gly-OH

A327689-10G A327689-1G A327689-250MG
76989-210EA 78 USD
76989-210 76989-212 76989-214
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)decanoic acid 95%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)decanoic acid

Precaución: for Research Use Only

Formula: C₂₅H₃₁NO₄
MW: 409.526 g/mol
MDL Number: MFCD01311753
CAS Number: 193885-59-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR