1-Benzylpyrrolidine-3-carbonitrile oxalate 97%

Supplier: Ambeed
Warning

A327614-1G A327614-5G
77032-838EA 53.05 USD
77032-838 77032-840
1-Benzylpyrrolidine-3-carbonitrile oxalate 97%
1-Benzylpyrrolidine-3-carbonitrile oxalate

Precaución: for Research Use Only

Formula: C₁₄H₁₆N₂O₄
MDL Number: MFCD11501670
CAS Number: 1188265-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR