3-Benzyl-1H-indazole 95%

Supplier: Ambeed
Warning

A327582-1G A327582-250MG
77032-834EA 92 USD
77032-834 77032-836
3-Benzyl-1H-indazole 95%
3-Benzyl-1H-indazole

Precaución: for Research Use Only

Formula: C₁₄H₁₂N₂
MDL Number: MFCD01565718
CAS Number: 4498-74-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR