(3R,4R)-tert-Butyl 3-fluoro-4-hydroxypiperidine-1-carboxylate 98%
Supplier: Ambeed
|
Warning
|
A327506-100MG
A327506-1G
A327506-250MG
77015-360EA
111.4
USD
77015-360
77015-362
77015-364
(3R,4R)-tert-Butyl 3-fluoro-4-hydroxypiperidine-1-carboxylate 98%
(3R,4R)-tert-Butyl 3-fluoro-4-hydroxypiperidine-1-carboxylate
Precaución: for Research Use Only
Formula:
C₁₀H₁₈FNO₃ MW: 219.25 g/mol |
MDL Number:
MFCD22123259 CAS Number: 1174020-43-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...