4-(4-Methyl-1-piperazinylcarbonyl)phenylboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 4-(4-Methyl-1-piperazinylcarbonyl)benzeneboronic acid pinacol ester, 4-(4-Methylpiperazine-1-carbonyl)phenylboronic acid, pinacol ester, (4-Methylpiperazin-1-yl)(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanone

A327414-250MG A327414-1G A327414-5G
77032-832EA 25 USD
77032-832 77601-478 77672-684
4-(4-Methyl-1-piperazinylcarbonyl)phenylboronic acid pinacol ester 97%
4-(4-Methyl-1-piperazinylcarbonyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₈H₂₇BN₂O₃
MDL Number: MFCD05863914
CAS Number: 832114-06-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR