2-Amino-4-(3-pyridyl)thiazole 97%

Supplier: Ambeed
Warning

Synonyms: 4-(Pyridin-3-yl)thiazol-2-amine

A327048-1G A327048-250MG
77015-358EA 78.51 USD
77015-358 77612-886
2-Amino-4-(3-pyridyl)thiazole 97%
2-Amino-4-(3-pyridyl)thiazole

Precaución: for Research Use Only

Formula: C₈H₇N₃S
MW: 177.22 g/mol
MDL Number: MFCD00218013
CAS Number: 30235-27-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR