cis-Octahydroisoindole 95%

Supplier: Ambeed
Warning

Synonyms: cis-Octahydro-1H-isoindole

A326922-10G A326922-1G A326922-25G A326922-5G
77002-972EA 65 USD
77002-972 77002-974 77002-976 77002-978
cis-Octahydroisoindole 95%
cis-Octahydroisoindole

Precaución: for Research Use Only

Formula: C₈H₁₅N
MDL Number: MFCD07778620
CAS Number: 1470-99-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR