3-(4-Bromophenyl)piperidin-2-one 95%

Supplier: Ambeed
Warning

A326806-100MG A326806-1G A326806-250MG
77015-352EA 45.62 USD
77015-352 77015-354 77015-356
3-(4-Bromophenyl)piperidin-2-one 95%
3-(4-Bromophenyl)piperidin-2-one

Precaución: for Research Use Only

Formula: C₁₁H₁₂BrNO
MW: 254.12 g/mol
MDL Number: MFCD17169922
CAS Number: 769944-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR