5-Methyl-1,3-benzoxazol-2-amine 95%

Supplier: Ambeed
Warning

Synonyms: 5-Methyl-2-aminobenzoxazol

A326793-1G A326793-250MG A326793-5G
77032-826EA 71.09 USD
77032-826 77032-828 77032-830
5-Methyl-1,3-benzoxazol-2-amine 95%
5-Methyl-1,3-benzoxazol-2-amine

Precaución: for Research Use Only

Formula: C₈H₈N₂O
MW: 148.162 g/mol
MDL Number: MFCD01664145
CAS Number: 64037-15-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR