tert-Butyl (1-(4-bromophenyl)cyclobutyl)carbamate 97%

Supplier: Ambeed
Warning

A326516-100MG A326516-1G A326516-250MG
77032-820EA 17 USD
77032-820 77032-822 77032-824
tert-Butyl (1-(4-bromophenyl)cyclobutyl)carbamate 97%
tert-Butyl (1-(4-bromophenyl)cyclobutyl)carbamate

Precaución: for Research Use Only

Formula: C₁₅H₂₀BrNO₂
MW: 326.23 g/mol
MDL Number: MFCD18072661
CAS Number: 1032350-06-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR