Methyl-2-(4-methoxy-3-nitrophenyl)acetate 98%

Supplier: Ambeed
Warning

A326399-250MG A326399-5G A326399-1G
77002-952EA 23 USD
77002-952 77002-954 77881-914
Methyl-2-(4-methoxy-3-nitrophenyl)acetate 98%
Methyl-2-(4-methoxy-3-nitrophenyl)acetate

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO₅
MDL Number: MFCD09754963
CAS Number: 34837-88-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR